(S)-ɑ-Methyl-4-nitrobenzylamine hydrochloride ≥99.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAN0543-1GEA 0 CNY
TCIAN0543-1G
(S)-ɑ-Methyl-4-nitrobenzylamine hydrochloride ≥99.0% (通过滴定分析)
(S)-alpha-Methyl-4-nitrobenzylamine hydrochloride
分子式: C₈H₁₀N₂O₂.HCl
分子量: 202,64 g/mol
MDL编号: MFCD00066312
CAS号: 132873-57-5

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Specific rotation [a]20/D -6.7 to -7.7 deg (C=1, 0.05mol/L NaOH sol.)
Solubility in Water faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR