(S)-(-)-N-(5-Nitro-2-pyridyl)alaninol ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAN0547-100MGEA 0 CNY
TCIAN0547-100MG
(S)-(-)-N-(5-Nitro-2-pyridyl)alaninol ≥98.0% (通过HPLC,滴定分析)
(S)-(-)-N-(5-Nitro-2-pyridyl)alaninol
分子式: C₈H₁₁N₃O₃
分子量: 197,19 g/mol
MDL编号: MFCD00145287
CAS号: 115416-52-9

现在订购

产品技术规格

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR