4-Nitro-N-phenylphthalimide ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAN0574-25GEA 0 CNY
TCIAN0574-25G
4-Nitro-N-phenylphthalimide ≥98.0% (通过GC)
4-Nitro-N-phenylphthalimide
分子式: C₁₄H₈N₂O₄
MDL编号: MFCD00191559
CAS号: 40392-27-6

现在订购

产品技术规格

Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR