1-(4-硝基苯基)-1H-吡咯 ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAN0584-1GEA
0
CNY
TCIAN0584-1G
1-(4-硝基苯基)-1H-吡咯 ≥98.0% (通过GC)
1-(4-硝基苯基)-1H-吡咯
分子式:
C₁₀H₈N₂O₂ 分子量: 188,1800 g/mol |
MDL编号:
MFCD00119340 CAS号: 4533-42-0 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...