4-Nitrophenyl-β-D-glucuronide ≥98.0% (通过HPLC) substrate for beta-Glucuronidase

供应商: TCI EUROPE

TCIAN0618-100MGEA 0 CNY
TCIAN0618-100MG
4-Nitrophenyl-β-D-glucuronide ≥98.0% (通过HPLC) substrate for beta-Glucuronidase
4-Nitrophenyl-beta-D-glucuronide
分子式: C₁₂H₁₃NO₉
分子量: 315,23 g/mol
存储条件: Refrigerator
MDL编号: MFCD00036210
CAS号: 10344-94-2
EINECS: 233-753-0

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -100 ~ -104 deg (C=1, EtOH)
Water max. 7.0 %
Free p-Nitrophenol max. 0.05 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR