3-Noradamantanecarboxylic acid ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAN0621-1GEA
0
CNY
TCIAN0621-1G
3-Noradamantanecarboxylic acid ≥98.0% (通过GC,滴定分析)
3-Noradamantanecarboxylic acid
分子式:
C₁₀H₁₄O₂ 分子量: 166,22 g/mol |
MDL编号:
MFCD00213463 CAS号: 16200-53-6 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 105.0 to 110.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...