4-Nitrocumene ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAN0639-25GEA 0 CNY
TCIAN0639-25G
4-Nitrocumene ≥98.0% (通过GC)
4-Nitrocumene
分子式: C₉H₁₁NO₂
分子量: 165,19 g/mol
沸点: 142°C
闪点: 126°C
MDL编号: MFCD00039746
CAS号: 1817-47-6
EINECS: 217-326-6

现在订购

产品技术规格

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR