1,2,9-Nonanetriol ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAN0652-5GEA 0 CNY
TCIAN0652-5G
1,2,9-Nonanetriol ≥97.0% (通过GC)
1,2,9-Nonanetriol
分子式: C₉H₂₀O₃
分子量: 176,26 g/mol
MDL编号: MFCD01861292
CAS号: 382631-42-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 55.0 to 58.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR