(1S,2S)-2-(Naphthalene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAN0714-100MGEA 1450 CNY
TCIAN0714-100MG
(1S,2S)-2-(Naphthalene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥98.0% (通过HPLC,滴定分析)
(1S,2S)-2-(Naphthalene-2,3-dicarboximido)cyclohexanecarboxylic acid
分子式: C₁₉H₁₇NO₄
CAS号: 642995-16-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +32.0 to +35.0 deg (C=0.3, THF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR