Allyl[3-(2-nitrobenzenesulphonamido)propyl]carbamate ≥98.0% (通过HPLC)
供应商: TCI EUROPE
TCIAN0717-1GEA
0
CNY
TCIAN0717-1G
Allyl[3-(2-nitrobenzenesulphonamido)propyl]carbamate ≥98.0% (通过HPLC)
Allyl[3-(2-nitrobenzenesulphonamido)propyl]carbamate
分子式:
C₁₃H₁₇N₃O₆S 分子量: 343,35 g/mol |
CAS号:
312283-45-7 |
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...