4-Nitrophenyl-β-D-fucopyranoside ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAN0774-100MGEA
900
CNY
TCIAN0774-100MG
4-Nitrophenyl-β-D-fucopyranoside ≥98.0% (通过GC)
4-Nitrophenyl-beta-D-fucopyranoside
分子式:
C₁₂H₁₅NO₇ |
CAS号:
1226-39-7 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 181.0 to 185.0 deg-C |
Specific rotation [a]20/D | -73.0 to -77.0 deg (C=1, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...