2-(4-Nitrophenoxy)naphthalene ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAN0807-1GEA
0
CNY
TCIAN0807-1G
2-(4-Nitrophenoxy)naphthalene ≥98.0% (通过GC)
2-(4-Nitrophenoxy)naphthalene
分子式:
C₁₆H₁₁NO₃ 分子量: 265,27 g/mol |
MDL编号:
MFCD00427410 CAS号: 71311-82-5 |
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...