2-Octadecyl-1,4-dimethoxybenzene

供应商: TCI EUROPE

TCIAO0217-5GEA 0 CNY
TCIAO0217-5G
2-Octadecyl-1,4-dimethoxybenzene
2-Octadecyl-1,4-dimethoxybenzene
分子式: C₂₆H₄₆O₂
分子量: 390,65 g/mol
MDL编号: MFCD00142955
CAS号: 88702-69-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 52.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR