2,3,7,8,12,13,17,18-八乙基-21H,23H-卟吩 ≥95.0% (以总氮计)
供应商: TCI EUROPE
Synonyms:
八乙基卟吩
TCIAO0234-100MGEA
600
CNY
TCIAO0234-100MG
TCIAO0234-1G
2,3,7,8,12,13,17,18-八乙基-21H,23H-卟吩 ≥95.0% (以总氮计)
2,3,7,8,12,13,17,18-八乙基-21H,23H-卟吩
分子式:
C₃₆H₄₆N₄ 分子量: 534,79 g/mol |
MDL编号:
MFCD00009627 CAS号: 2683-82-1 EINECS: 220-243-8 |
产品技术规格
Appearance | Red to Dark blue to Black powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...