4-n-Octylstyrene ≥95.0% (通过GC) stabilised

供应商: TCI EUROPE


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

TCIAO0273-5GEA 0 CNY
TCIAO0273-5G
4-n-Octylstyrene ≥95.0% (通过GC) stabilised
4-n-Octylstyrene
分子式: C₁₆H₂₄
分子量: 216,37 g/mol
沸点: 114°C
存储条件: Refrigerator
MDL编号: MFCD00078353
CAS号: 46745-66-8

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR