ɑ-Octithiophene
供应商: TCI EUROPE
TCIAO0313-100MGEA
4975
CNY
TCIAO0313-100MG
ɑ-Octithiophene
alpha-Octithiophene
分子式:
C₃₂H₁₈S₈ 溶点: 364°C |
CAS号:
113728-71-5 |
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
TOF-MS | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...