2-n-Octyl-4-isothiazolin-3-one ≥98.0% (通过GC) for biochemical research

供应商: TCI EUROPE
Danger

TCIAO0378-1GEA 0 CNY
TCIAO0378-1G
2-n-Octyl-4-isothiazolin-3-one ≥98.0% (通过GC) for biochemical research
2-n-Octyl-4-isothiazolin-3-one
分子式: C₁₁H₁₉NOS
分子量: 213,34 g/mol
沸点: 142°C
MDL编号: MFCD00072473
CAS号: 26530-20-1
EINECS: 247-761-7
UN号: 2923
ADR: 8,II

现在订购

产品技术规格

Purity (GC) min. 98.0 %
Protein impurity test (SDS-PAGE) to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR