5-Propylhydantoin ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE

TCIAP0323-25GEA 0 CNY
TCIAP0323-25G
5-Propylhydantoin ≥98.0% (通过GC,滴定分析)
5-Propylhydantoin
分子式: C₆H₁₀N₂O₂
分子量: 142,16 g/mol
MDL编号: MFCD00059721
CAS号: 18227-41-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 137.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR