3-吡啶丙醇 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: 3-(3-吡啶基)-1-丙醇

TCIAP0350-10GEA 810 CNY
TCIAP0350-10G
3-吡啶丙醇 ≥98.0% (通过GC)
3-吡啶丙醇
分子式: C₈H₁₁NO
分子量: 137,18 g/mol
沸点: 133°C
MDL编号: MFCD00006414
CAS号: 2859-67-8
EINECS: 220-675-7

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR