2-(4-吡啶基)乙磺酸 ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

Synonyms: 4-吡啶乙磺酸

TCIAP0353-25GEA 0 CNY
TCIAP0353-25G
2-(4-吡啶基)乙磺酸 ≥98.0% (通过滴定分析)
2-(4-吡啶基)乙磺酸
分子式: C₇H₉NO₃S
分子量: 187,21 g/mol
MDL编号: MFCD00006452
CAS号: 53054-76-5
EINECS: 258-331-3
UN号: 2585
ADR: 8,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR