Butyl palmitate ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAP0649-25MLEA 0 CNY
TCIAP0649-25ML
Butyl palmitate ≥95.0% (通过GC)
Butyl palmitate
分子式: C₂₀H₄₀O₂
分子量: 312,54 g/mol
沸点: 170°C
MDL编号: MFCD00053738
CAS号: 111-06-8
EINECS: 203-829-8

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR