Bis(trans-3,3,5-trimethylcyclohexyl)phthalate ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAP0697-25GEA 0 CNY
TCIAP0697-25G
Bis(trans-3,3,5-trimethylcyclohexyl)phthalate ≥95.0% (通过GC)
Bis(trans-3,3,5-trimethylcyclohexyl)phthalate
分子式: C₂₆H₃₈O₄
分子量: 414,59 g/mol
MDL编号: MFCD00070481
CAS号: 245652-82-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 50.0 to 57.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR