2-Propylvaleryl chloride ≥99.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAP0824-5MLEA 0 CNY
TCIAP0824-5ML
2-Propylvaleryl chloride ≥99.0% (通过GC)
2-Propylvaleryl chloride
分子式: C₈H₁₅ClO
分子量: 162,66 g/mol
沸点: 132°C
MDL编号: MFCD00051446
CAS号: 2936-08-5
EINECS: 220-914-5
UN号: 3265
ADR: 8,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR