4-Hexyloxyphenyl-4-pentylbenzoate ≥99.0% (通过GC)

供应商: TCI EUROPE

TCIAP0897-1GEA 0 CNY
TCIAP0897-1G
4-Hexyloxyphenyl-4-pentylbenzoate ≥99.0% (通过GC)
4-Hexyloxyphenyl-4-pentylbenzoate
分子式: C₂₄H₃₂O₃
分子量: 368,52 g/mol
MDL编号: MFCD00059437
CAS号: 50802-52-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR