3-Pentenenitrile ≥95.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAP0958-25MLEA 0 CNY
TCIAP0958-25ML
3-Pentenenitrile ≥95.0% (通过GC)
3-Pentenenitrile
分子式: C₅H₇N
分子量: 81,12 g/mol
沸点: 144°C
闪点: 40°C
MDL编号: MFCD00001963
CAS号: 4635-87-4
EINECS: 225-060-7
UN号: 3275
ADR: 6.1,II

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR