3,4,9,10-Perylenetetracarboxylic diimide ≥95.0% (以总氮计)

TCIAP0984-25GEA 510 CNY
TCIAP0984-25G
3,4,9,10-Perylenetetracarboxylic diimide ≥95.0% (以总氮计)
3,4,9,10-Perylenetetracarboxylic diimide
分子式: C₂₄H₁₀N₂O₄
分子量: 390,35 g/mol
MDL编号: MFCD00024144
CAS号: 81-33-4
EINECS: 201-344-6

现在订购

产品技术规格

Appearance Red to Dark blue to Black powder to crystal
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR