1-胡椒基哌嗪 ≥97.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAP1041-25GEA
0
CNY
TCIAP1041-25G
1-胡椒基哌嗪 ≥97.0% (通过滴定分析)
1-胡椒基哌嗪
分子式:
C₁₂H₁₆N₂O₂ 分子量: 220,27 g/mol 沸点: 149°C |
MDL编号:
MFCD00005834 CAS号: 32231-06-4 EINECS: 250-968-5 |
产品技术规格
Appearance | White to Yellow to Orange powder to crystaline |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 40.0 to 46.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...