1-(10,12-Pentacosadiynyl)pyridinium bromide

供应商: TCI EUROPE
Warning

TCIAP1054-100MGEA 2350 CNY
TCIAP1054-100MG
1-(10,12-Pentacosadiynyl)pyridinium bromide
1-(10,12-Pentacosadiynyl)pyridinium bromide
分子式: C₃₀H₄₈BrN
存储条件: Freezer
MDL编号: MFCD00059097
CAS号: 94598-31-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Elemental analysis (Nitrogen) 2.65 to 3.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR