2-Phenylcarbamoyl-1,4-naphthoquinone-4-(4-diethylamino-2-methylphenyl)imine ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
TCIAP1149-5GEA
0
CNY
TCIAP1149-5G
2-Phenylcarbamoyl-1,4-naphthoquinone-4-(4-diethylamino-2-methylphenyl)imine ≥98.0% (通过滴定分析)
2-Phenylcarbamoyl-1,4-naphthoquinone-4-(4-diethylamino-2-methylphenyl)imine
分子式:
C₂₈H₂₇N₃O₂ 分子量: 437,54 g/mol |
MDL编号:
MFCD00191571 CAS号: 102187-19-9 |
产品技术规格
Appearance | Green to Dark green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in THF | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...