4-Propionylbiphenyl ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAP1212-5GEA 0 CNY
TCIAP1212-5G
4-Propionylbiphenyl ≥98.0% (通过GC)
4-Propionylbiphenyl
分子式: C₁₅H₁₄O
分子量: 210,28 g/mol
沸点: 344°C
MDL编号: MFCD00027015
CAS号: 37940-57-1

现在订购

产品技术规格

Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR