(R)-(-)-ɑ-(Trifluoromethyl)benzyl alcohol

供应商: TCI EUROPE
Warning

Synonyms: (R)-(-)-1-苯基-2,2,2-三氟乙醇(-)-苯基(三氟甲基)甲醇

TCIAP1367-1GEA 0 CNY
TCIAP1367-1G
(R)-(-)-ɑ-(Trifluoromethyl)benzyl alcohol
(R)-(-)-α-(三氟甲基)苄醇
分子式: C₈H₇F₃O
分子量: 176,14 g/mol
沸点: 99°C
闪点: 87°C
MDL编号: MFCD00077844
CAS号: 10531-50-7
EINECS: 234-094-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -30.0 to -33.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR