(S)-(+)-α-(三氟甲基)苄醇 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
(+)-苯基(三氟甲基)甲醇
, (S)-(+)-1-苯基-2,2,2-三氟乙醇
TCIAP1368-1GEA
0
CNY
TCIAP1368-1G
(S)-(+)-α-(三氟甲基)苄醇 ≥98.0% (通过GC)
(S)-(+)-α-(三氟甲基)苄醇
分子式:
C₈H₇F₃O 分子量: 176,14 g/mol 沸点: 127°C 闪点: 84°C |
MDL编号:
MFCD00077845 CAS号: 340-06-7 EINECS: 206-430-7 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +30.0 to +33.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...