1-Naphthyl propionate

供应商: TCI EUROPE

Synonyms: 丙酸-1-萘酯

TCIAP1384-5GEA 0 CNY
TCIAP1384-5G
1-Naphthyl propionate
1-萘基丙酸酯
分子式: C₁₃H₁₂O₂
分子量: 200,24 g/mol
沸点: 135°C
MDL编号: MFCD00010496
CAS号: 3121-71-9

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR