2-(Phenylsulphonylmethyl)benzaldehyde ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAP1455-1GEA 0 CNY
TCIAP1455-1G
2-(Phenylsulphonylmethyl)benzaldehyde ≥97.0% (通过GC)
2-(Phenylsulphonylmethyl)benzaldehyde
分子式: C₁₄H₁₂O₃S
分子量: 260,31 g/mol
MDL编号: MFCD06797157
CAS号: 468751-38-4

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 141.0 to 145.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR