2-(Phenylsulphonylmethyl)benzonitrile ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAP1456-1GEA 0 CNY
TCIAP1456-1G
2-(Phenylsulphonylmethyl)benzonitrile ≥98.0% (通过GC)
2-(Phenylsulphonylmethyl)benzonitrile
分子式: C₁₄H₁₁NO₂S
分子量: 257,31 g/mol
MDL编号: MFCD06797158
CAS号: 82651-72-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR