1,2,3-Propanetricarboxamide ≥98.0% (以总氮计)
供应商: TCI EUROPE
TCIAP1732-1GEA
0
CNY
TCIAP1732-1G
1,2,3-Propanetricarboxamide ≥98.0% (以总氮计)
1,2,3-Propanetricarboxamide
分子式:
C₆H₁₁N₃O₃ 分子量: 173,17 g/mol 溶点: 218°C |
CAS号:
1205674-38-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...