1,2,3,4,6-Penta-O-pivaloyl-D-mannopyranose ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAP1803-1GEA
0
CNY
TCIAP1803-1G
1,2,3,4,6-Penta-O-pivaloyl-D-mannopyranose ≥98.0% (通过GC)
1,2,3,4,6-Penta-O-pivaloyl-D-mannopyranose
分子式:
C₃₁H₅₂O₁₁ 分子量: 600,75 g/mol 存储条件: Refrigerator |
CAS号:
220017-47-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Specific rotation [a]20/D | +37.0 to +43.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...