2-吡啶基三氟甲磺酸酯 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: 2-吡啶基三氟甲基磺酸酯

TCIAP1840-5GEA 0 CNY
TCIAP1840-5G
2-吡啶基三氟甲磺酸酯 ≥98.0% (通过GC)
2-吡啶基三氟甲磺酸酯
分子式: C₆H₄F₃NO₃S
分子量: 227,16 g/mol
沸点: 108°C
闪点: 27°C
MDL编号: MFCD00192531
CAS号: 65007-00-3
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR