5-Phenoxyamyl bromide ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAP1847-5GEA 845 CNY
TCIAP1847-5G
5-Phenoxyamyl bromide ≥95.0% (通过GC)
5-Phenoxyamyl bromide
分子式: C₁₁H₁₅BrO
沸点: 130°C
MDL编号: MFCD00039203
CAS号: 22921-72-8

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 1.2780 to 1.2820
Refractive index n20/D 1.5330 to 1.5370

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR