6-Phenoxyhexyl bromide ≥96.0% (通过GC)

供应商: TCI EUROPE

TCIAP1848-5GEA 0 CNY
TCIAP1848-5G
6-Phenoxyhexyl bromide ≥96.0% (通过GC)
6-Phenoxyhexyl bromide
分子式: C₁₂H₁₇BrO
分子量: 257,17 g/mol
沸点: 112°C
MDL编号: MFCD00039205
CAS号: 51795-97-2

现在订购

产品技术规格

Appearance White to Almost white clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR