Pyrimidine-5-carboxylic acid ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAP1909-1GEA 0 CNY
TCIAP1909-1G
Pyrimidine-5-carboxylic acid ≥97.0% (通过GC,滴定分析)
Pyrimidine-5-carboxylic acid
分子式: C5H4N2O2
分子量: 124,10 g/mol
溶点: 270°C
MDL编号: MFCD00856162
CAS号: 4595-61-3
EINECS: 224-994-2

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR