(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAP1925-1GEA 0 CNY
TCIAP1925-1G
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine ≥98.0% (通过GC,滴定分析)
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine
分子式: C₉H₁₈N₂
分子量: 154,2500 g/mol
MDL编号: MFCD04117879
CAS号: 60419-23-0

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -7.0 to -11.0 deg (C=2.4, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR