奎尼丁 ≥85.0% (通过HPLC)
供应商: TCI EUROPE
|
Danger
|
TCIAQ0006-5GEA
0
CNY
TCIAQ0006-5G
TCIAQ0006-25G
奎尼丁 ≥85.0% (通过HPLC)
奎尼丁
分子式:
C20H24N2O2 分子量: 324,42 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00135581 CAS号: 56-54-2 EINECS: 200-279-0 UN号: 2811 ADR: 6.1,III |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 85.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 169.0 to 174.0 deg-C |
Specific rotation [a]20/D | +260 to +275 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...