Calcium(2E,4E)hexa-2,4-dienoate ≥95.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAS0054-25GEA 320 CNY
TCIAS0054-25G
Calcium(2E,4E)hexa-2,4-dienoate ≥95.0% (通过滴定分析)
Calcium(2E,4E)hexa-2,4-dienoate
分子式: C₁₂H₁₄CaO₄
分子量: 262,3200 g/mol
MDL编号: MFCD00043805
CAS号: 7492-55-9
EINECS: 231-321-6

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Chelometric Titration) min. 95.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR