2-Carboxyphenyl salicylate ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAS0129-25GEA 0 CNY
TCIAS0129-25G TCIAS0129-100G
2-Carboxyphenyl salicylate ≥98.0% (通过HPLC,滴定分析)
2-Carboxyphenyl salicylate
分子式: C₁₄H₁₀O₅
分子量: 258,23 g/mol
MDL编号: MFCD00020252
CAS号: 552-94-3
EINECS: 209-027-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 135.0 to 144.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR