Bis(2-ethylhexyl)suberate ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAS0443-25MLEA 0 CNY
TCIAS0443-25ML
Bis(2-ethylhexyl)suberate ≥98.0% (通过GC)
Bis(2-ethylhexyl)suberate
分子式: C₂₄H₄₆O₄
分子量: 398,63 g/mol
MDL编号: MFCD01631311
CAS号: 5238-22-2

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR