1,3,3-Trimethylindolino-6'-bromobenzopyrylospiran ≥98.0% (通过HPLC,滴定分析) photochromic compound

供应商: TCI EUROPE

TCIAT0370-1GEA 0 CNY
TCIAT0370-1G
1,3,3-Trimethylindolino-6'-bromobenzopyrylospiran ≥98.0% (通过HPLC,滴定分析) photochromic compound
1,3,3-Trimethylindolino-6'-bromobenzopyrylospiran
分子式: C₁₉H₁₈BrNO
分子量: 356,26 g/mol
MDL编号: MFCD00059896
CAS号: 16650-14-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR