2,4,5-Trichlorophenol sodium salt ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAT0391-25GEA 360 CNY
TCIAT0391-25G
2,4,5-Trichlorophenol sodium salt ≥98.0% (通过GC)
2,4,5-Trichlorophenol sodium salt
分子式: C₆H₃Cl₃O
MDL编号: MFCD00058974
CAS号: 136-32-3
EINECS: 205-239-6
UN号: 2811
ADR: 6.1,III

现在订购

产品技术规格

Appearance Light orange to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR