2,4,6-Triphenylnitrobenzene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAT0518-10GEA 0 CNY
TCIAT0518-10G
2,4,6-Triphenylnitrobenzene ≥98.0% (通过GC)
2,4,6-Triphenylnitrobenzene
分子式: C₂₄H₁₇NO₂
MDL编号: MFCD00060090
CAS号: 10368-47-5

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR