2,4,4-Trimethyl-1-hexene ≥97.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAT0778-5MLEA 0 CNY
TCIAT0778-5ML
2,4,4-Trimethyl-1-hexene ≥97.0% (通过GC)
2,4,4-Trimethyl-1-hexene
分子式: C₉H₁₈
分子量: 126,24 g/mol
沸点: 132°C
闪点: 24°C
MDL编号: MFCD00048626
CAS号: 51174-12-0
UN号: 3295
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR